Få mere at vide
D-Pemetrexed Dimethyl Diester, TRC
Artikelnummer. 30416985
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416985 100 mg
30423466 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30416985 Leverandør Toronto Research Chemicals Leverandørnr. TRCP219535100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1391068-12-4
Molekylær formel C22H25N5O6
Molekylvægt (g/mol) 455.46
Synonym N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid 1,5-Dimethyl Diester
IUPAC navn dimethyl (2R)-2-[[4-[2-(2-amino-4-oxo-1,7-dihydropyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]amino]pentanedioate
SMIL COC(=O)CC[C@@H](NC(=O)c1ccc(CCc2c[nH]c3NC(=NC(=O)c23)N)cc1)C(=O)OC
Tekniske data
Kemisk navn eller materiale D-Pemetrexed Dimethyl Diester
Mængde 100 mg
InChI formel InChI=1S/C22H25N5O6/c1-32-16(28)10-9-15(21(31)33-2)25-19(29)13-6-3-12(4-7-13)5-8-14-11-24-18-17(14)20(30)27-22(23)26-18/h3-4,6-7,11,15H,5,8-10H2,1-2H3,(H,25,29)(H4,23,24,26,27,30)/t15-/m1/s1
Formel vægt 455.1805