Få mere at vide
D-Pantethine, TRC
Artikelnummer. 30408987
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408987 50 mg
30420283 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30408987 Leverandør Toronto Research Chemicals Leverandørnr. TRCP20582350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Pantethine
CAS 16816-67-4
Molekylær formel C22 H42 N4 O8 S2
Mængde 50 mg
Synonym Butanamide','N,N'-[dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethyl-','(2R,2'R)-, Butanamide','N,N'-[dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethyl-','[R-(R*,R*)]-, Butyramide','N,N'-[dithiobis(ethyleneiminocarbonylethylene)]bis[2,4-dihydroxy-3,3-dimethyl-','D-(+)- (8CI), (2R,2'R)-N,N'-[Dithiobis[2,1-ethanediylimino(3-oxo-3,1-propanediyl)]]bis[2,4-dihydroxy-3,3-dimethylbutanamide], Bis(N-pantothenylamidoethyl) disulfide, Bis(pantothenamidoethyl) disulfide, D-Bis-pantothenyl-β-aminoethyl disulfide, D-Pantethine, LBF disulfide, Lipodel, Pantethine, Pantetina, Panthecin, Pantomin, Pantosin
InChI formel InChI=1S/C22H42N4O8S2/c1-21(2,13-27)17(31)19(33)25-7-5-15(29)23-9-11-35-36-12-10-24-16(30)6-8-26-20(34)18(32)22(3,4)14-28/h17-18,27-28,31-32H,5-14H2,1-4H3,(H,23,29)(H,24,30)(H,25,33)(H,26,34)/t17-,18-/m0/s1
SMIL CC(C)(CO)[C@@H](O)C(=O)NCCC(=O)NCCSSCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)CO
IUPAC navn (2R)-N-[3-[2-[2-[3-[[(2R)-2,4-dihydroxy-3,3-dimethylbutanoyl]amino]propanoylamino]ethyldisulfanyl]ethylamino]-3-oxopropyl]-2,4-dihydroxy-3,3-dimethylbutanamide
Molekylvægt (g/mol) 554.72
Formel vægt 554.2444