Få mere at vide
D-(+)-Noradrenaline Tartrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | D-(+)-Noradrenaline Tartrate |
| CAS | 636-88-4 |
| Analyt- eller komponentnavne | D-(+)-Noradrenaline Tartrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C8 H11 N O3 . C4 H6 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,2-Benzenediol, 4-[(1S)-2-amino-1-hydroxyethyl]-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), 1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, (S)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), 1,2-Benzenediol, 4-[(1S)-2-amino-1-hydroxyethyl]-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt), Benzyl alcohol, α-(aminomethyl)-3,4-dihydroxy-, (+)-, tartrate (1:1) (salt), (+)-(S)-Norepinephrine bitartrate, (+)-Noradrenaline (+)-bitartrate, (+)-Noradrenaline bitartrate, (+)-Norepinephrine (+)-bitartrate, (+)-Norepinephrine bitartrate, D-Norepinephrine bitartrate, D-Norepinephrine d-bitartrate, S-(+)-Norepinephrine bitartrate, d-Noradrenaline bitartrate, d-Noradrenaline hydrogen tartrate |
| InChI formel | InChI=1S/C8H11NO3.C4H6O6/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10)/t8-;1-,2-/m11/s1 |
| Løsningstype | Neat |
| SMIL | NC[C@@H](O)c1ccc(O)c(O)c1.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| Vis mere |