Få mere at vide
D-Neopterin-13C5, TRC
Artikelnummer. 30009599
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009599 10 mg
30005627 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009599 Leverandør Toronto Research Chemicals Leverandørnr. TRCN39000710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1217632-04-6
Molekylær formel C413C5H11N5O4
Molekylvægt (g/mol) 258.18
Synonym 2-Amino-6-[(1S,2R)-1,2,3-trihydroxypropyl]-4(3H)-pteridinone-13C5, Neopterin-13C5, 2-Amino-4-hydroxy-6-(1,2,3-trihydroxypropyl)pteridine-13C5, 2-Amino-6-((1S,2R)-1,2,3-trihydroxypropyl)-4(8H)-pteridone-13C5, 6-D-erythro-Neopterin-13C5, D-(+)-Neopterin-13C5, D-erythro-Neopterin-13C5;
SMIL O=C1C2=N[13C]([13C@H](O)[13C@H](O)[13CH2]O)=[13CH]NC2=NC(N)=N1
Tekniske data
Kemisk navn eller materiale D-Neopterin-13C5
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C9H11N5O4/c10-9-13-7-5(8(18)14-9)12-3(1-11-7)6(17)4(16)2-15/h1,4,6,15-17H,2H2,(H3,10,11,13,14,18)
Formel vægt 258.0979
Procent renhed >95%