Learn More
Alfa Aesar™ D-myo-Inositol-1,2-bisphosphate sodium salt, 98%
Brand: Alfa Aesar™ J66420.|Z
Additional Details : Weight : 0.00001kg
Quantity | 0.1mg |
---|
Chemical Identifiers
208584-51-4 | |
386.093 | |
PHZDHEPQFXTBRQ-JBGYRJLXSA-N | |
131875843 | |
C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na] |
C6H14Na2O12P2 | |
MFCD10567083 | |
d-myo-inositol-1,2-diphosphate sodium salt | |
sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate |
Specifications
D-myo-Inositol-1,2-bisphosphate sodium salt | |
0.1mg | |
C6H14Na2O12P2 | |
d-myo-inositol-1,2-diphosphate sodium salt | |
C1(C(C(C(C(C1O)OP(=O)(O)O)OP(=O)(O)O)O)O)O.[Na].[Na] | |
386.093 | |
384.08 | |
98% |
White | |
208584-51-4 | |
MFCD10567083 | |
PHZDHEPQFXTBRQ-JBGYRJLXSA-N | |
sodium;[(1S,2R,3S,4S,5R,6R)-2,3,4,5-tetrahydroxy-6-phosphonooxycyclohexyl] dihydrogen phosphate | |
131875843 | |
Lyophilized Powder |