Få mere at vide
D-Lyxonic Acid, Potassium Salt, TRC
Artikelnummer. 30482115
Skift visning
Mængde:
250 mg
1 g
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30482115 2.5 g 2.5g
30465276 250 mg -
30464532 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30482115 Leverandør Toronto Research Chemicals Leverandørnr. TRCL4899502.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 78138-87-1
Molekylær formel C5H9KO6
Molekylvægt (g/mol) 204.22
IUPAC navn potassium;(2S,3S,4R)-2,3,4,5-tetrahydroxypentanoate
SMIL [K+].OC[C@@H](O)[C@H](O)[C@H](O)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale D-Lyxonic Acid, Potassium Salt
Mængde 2.5 g
InChI formel InChI=1S/C5H10O6.K/c6-1-2(7)3(8)4(9)5(10)11;/h2-4,6-9H,1H2,(H,10,11);/q;+1/p-1/t2-,3+,4+;/m1./s1
Formel vægt 204.0036