Learn More
D-Lysine Methyl Ester Dihydrochloride, TRC
Product Code. 30470893
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity Unit Size
30470893 5 g 5g
1 options
Product Code. 30470893 Supplier Toronto Research Chemicals Supplier No. TRCL4689205G
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material D-Lysine Methyl Ester Dihydrochloride
CAS 67396-08-1
Molecular Formula C7 H16 N2 O2 . 2[HCl]
Quantity 5 g
InChI Formula InChI=1S/C7H16N2O2.2ClH/c1-11-7(10)6(9)4-2-3-5-8;;/h6H,2-5,8-9H2,1H3;2*1H/t6-;;/m1../s1
SMILES Cl.Cl.COC(=O)[C@H](N)CCCCN
IUPAC Name methyl (2R)-2,6-diaminohexanoate;dihydrochloride
Molecular Weight (g/mol) 233.136
Formula Weight 232.075