Learn More
Abcam D-Luciferin potassium salt, chemiluminescent luciferase substrate
Product Code. 32119869 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
32119869 1 g 1g
1 options
Product Code. 32119869 Supplier Abcam Supplier No. ab1436551gsld
 Restricted Item
Forventet forsendelse: 19-11-2026
MW 318.4 Da, Purity >98%. Chemiluminescent luciferase substrate. Water soluble derivative of D-Luciferin (ab143654) and analog of D-Luciferin sodium salt (ab145164). Emits light on oxidative decarboxylation in the presence of ATP providing a bioluminescent signal for cellular ATP levels.
MW 318.4 Da, Purity >98%. Chemiluminescent luciferase substrate. Water soluble derivative of D-Luciferin (ab143654) and analog of D-Luciferin sodium salt (ab145164). Emits light on oxidative decarboxylation in the presence of ATP providing a bioluminescent signal for cellular ATP levels in vivo..
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Form Solid
Quantity 1 g
Regulatory Status RUO