Få mere at vide
D-Leucine-d10-N-FMOC, CDN
Artikelnummer. 30311288
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311288 0.05 g -
30310745 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311288 Leverandør CDN Leverandørnr. CDND79780.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-leucine-2,3,3,4,5,5,5,5',5',5'-D10
CAS 2714483-30-2
Molekylær formel C21 D10 H13 N O4
Mængde 0.05 g
Synonym N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-leucine-2,3,3,4,5,5,5,5',5',5'-d10, D-Leucine-d10-N-FMOC, (2 R)-2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]-4-(methyl-d3)pentanoic-2,3,3,4,5,5,5-d7 acid, N-9-Fluorenylmethoxycarbonyl-D-leucine-d10, N-FMOC-D-leucine-2,3,3,4,5,5,5,5',5',5'-d10, N-FMOC-D-leucine-2,3,3,4,5,5,5,5',5',5'-d10
InChI formel InChI=1 S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19 H,11-12H2,1-2H3,(H,22,25)(H,23,24)/t19-/m1/s1/i1D3,2D3,11D2,13 D,19 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[C@@]([2 H])(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O
IUPAC navn (2 R)-2,3,3,4,5,5,5-heptadeuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-4-(trideuteriomethyl)pentanoic acid
Molekylvægt (g/mol) 363.4732
Formel vægt 363.2255 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity