missing translation for 'onlineSavingsMsg'
Learn More

β-D-Lactose, ca 80% β and 20% α, ACROS Organics™

Brand:  Acros Organics 412980020

Additional Details : CAS Number : 5965-66-2 Weight : 2.05000kg

Packaging Plastic bottle
Quantity 2kg

 View more versions of this product

Product Code. 10475651

  • 920.00 DKK / 2kg

In stock, ready to ship: 12

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

5965-66-2
342.297
GUBGYTABKSRVRQ-DCSYEGIMSA-N
6134
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
C12H22O11
MFCD00064521
beta-d-lactose, beta-lactose, lactose, lactose, anhydrous, fast-flo lactose, milk sugar, d-lactose, 4-o-beta-d-galactopyranosyl-beta-d-glucopyranose, fast-flo, lactose anhydrous
CHEBI:36218
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O

Specifications

Passes Test (per Eur. Pharm.)
Passes Test (per Eur. Pharm.)
Authentic
Plastic bottle
On anhydrous basis: +54.4° to +55.9° (20°C, 589nm) (c=10, H2O/NH3)
1.0% max. (K.F.)
2kg
C12H22O11
beta-d-lactose, beta-lactose, lactose, lactose, anhydrous, fast-flo lactose, milk sugar, d-lactose, 4-o-beta-d-galactopyranosyl-beta-d-glucopyranose, fast-flo, lactose anhydrous
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O
342.297
CHEBI:36218
Crystalline Powder
0.4mL 0.1N NaOH/6g max.
β-D-Lactose
0.5% max. (1 g, 80°C, 2 hrs)
+55.15° (20°C c=10,water,NH3)
0.1% max.
White
5965-66-2
MFCD00064521
GUBGYTABKSRVRQ-DCSYEGIMSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
6134
342.29
≥78% (beta-lactose), 22% max. (alpha-lactose)
Videos
SDS Documents