Få mere at vide
D,L-Tryptophan-d3, TRC
Artikelnummer. 30450394
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30450394 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450394 Leverandør Toronto Research Chemicals Leverandørnr. TRCT94720225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale DL-Tryptophan-d3
CAS 340257-61-6
Molekylær formel C11 2H3 H9 N2 O2
Mængde 25 mg
Synonym Tryptophan-α,β,β-d3 (9CI), D,L-Tryptophan-d3, DL-Tryptophan-d3, Tryptophan-d3, α-Amino-3-indole-α,β,β-d3-propionic acid, α-Aminoindole-3-propanoic-d3 acid, Tryptophan-α,β,β-D3
InChI formel InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/i5D2,9D
SMIL [2H]C([2H])(c1c[nH]c2ccccc12)C([2H])(N)C(=O)O
IUPAC navn 2-amino-2,3,3-trideuterio-3-(1H-indol-3-yl)propanoic acid
Molekylvægt (g/mol) 207.24
Formel vægt 207.1087