Få mere at vide
D,L-Sulforaphane-L-cysteine, TRC
Artikelnummer. 30138511
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138511 10 mg
30134482 100 mg
17959524 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138511 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69941010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 364083-21-6
Molekylær formel C9 H18 N2 O3 S3
Molekylvægt (g/mol) 298.45
IUPAC navn (2R)-2-amino-3-(4-methylsulfinylbutylcarbamothioylsulfanyl)propanoic acid
SMIL CS(=O)CCCCNC(=S)SC[C@H](N)C(=O)O
Tekniske data
Kemisk navn eller materiale D,L-Sulforaphane-L-cysteine
Renhedsgrad noter HPLC
InChI formel InChI=1S/C9H18N2O3S3/c1-17(14)5-3-2-4-11-9(15)16-6-7(10)8(12)13/h7H,2-6,10H2,1H3,(H,11,15)(H,12,13)/t7-,17?/m0/s1
Mængde 10 mg
Formel vægt 298.048
Procent renhed >95%