missing translation for 'onlineSavingsMsg'
Få mere at vide

D,L-Mevalonic Acid Lactone-d3, TRC

Artikelnummer. 30480034
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480034 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30480034 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3390271MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale D,L-Mevalonic Acid Lactone-d3
CAS 61219-76-9
Molekylær formel C6 2H3 H7 O3
Mængde 1 mg
Synonym 2H-Pyran-2-one, tetrahydro-4-hydroxy-4-(methyl-d3)-, Tetrahydro-4-hydroxy-4-(methyl-d3)-2H-pyran-2-one, (±)-Mevalolactone-d3, (±)-Mevalonolactone-d3, DL-Mevalonic acid lactone-d3, DL-Mevalonic acid Δ-lactone-d3, DL-Mevalonolactone-d3, β-Hydroxy-β-(methyl-d3)-Δ-valerolactone, (±)-Mevalolactone-D3 (methyl-D3)
InChI formel InChI=1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3
SMIL [2H]C([2H])([2H])C1(O)CCOC(=O)C1
IUPAC navn 4-hydroxy-4-(trideuteriomethyl)oxan-2-one
Molekylvægt (g/mol) 133.16
Formel vægt 133.0818
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.