Få mere at vide
D,L-Mevalonic Acid Lactone-d3, TRC
Artikelnummer. 30474098
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30474098 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30474098 Leverandør Toronto Research Chemicals Leverandørnr. TRCM33902710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D,L-Mevalonic Acid Lactone-d3
CAS 61219-76-9
Molekylær formel C6 2H3 H7 O3
Mængde 10 mg
Synonym 2H-Pyran-2-one, tetrahydro-4-hydroxy-4-(methyl-d3)-, Tetrahydro-4-hydroxy-4-(methyl-d3)-2H-pyran-2-one, (±)-Mevalolactone-d3, (±)-Mevalonolactone-d3, DL-Mevalonic acid lactone-d3, DL-Mevalonic acid Δ-lactone-d3, DL-Mevalonolactone-d3, β-Hydroxy-β-(methyl-d3)-Δ-valerolactone, (±)-Mevalolactone-D3 (methyl-D3)
InChI formel InChI=1S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3
SMIL [2H]C([2H])([2H])C1(O)CCOC(=O)C1
IUPAC navn 4-hydroxy-4-(trideuteriomethyl)oxan-2-one
Molekylvægt (g/mol) 133.16
Formel vægt 133.0818