Få mere at vide
D,L-Mevalonic Acid Dicyclohexylammonium Salt, TRC
Artikelnummer. 30480033
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480033 100 mg
30472495 50 mg
30472496 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480033 Leverandør Toronto Research Chemicals Leverandørnr. TRCM339020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1215802-31-5
Molekylær formel C18H35NO4
Molekylvægt (g/mol) 329.47
Synonym 3,5-Dihydroxy-3-(methyl) Pentanoic Acid Dicyclohexylammonium Salt, Hiochic Acid Dicyclohexylammonium Salt, 3,5-Dihydroxy-3-(methyl)valeric Acid Dicyclohexylammonium Salt, MK 91, MVA, MVS, (RS)-Mevalonic Acid Dicyclohexylammonium Salt
IUPAC navn N-cyclohexylcyclohexanamine;3,5-dihydroxy-3-methylpentanoic acid
SMIL CC(O)(CCO)CC(=O)O.C1CCC(CC1)NC2CCCCC2
Tekniske data
Kemisk navn eller materiale D,L-Mevalonic Acid Dicyclohexylammonium Salt
Mængde 100 mg
InChI formel InChI=1S/C12H23N.C6H12O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(10,2-3-7)4-5(8)9/h11-13H,1-10H2;7,10H,2-4H2,1H3,(H,8,9)
Formel vægt 329.2566