Få mere at vide
D,L-Gabaculine, Hydrochloride, TRC
Artikelnummer. 30137273
Skift visning
Mængde:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137273 250 mg
30135598 10 mg
30007347 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137273 Leverandør Toronto Research Chemicals Leverandørnr. TRCG111000250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59556-17-1
Molekylær formel C7 H9 N O2 . Cl H
Molekylvægt (g/mol) 175.61
IUPAC navn 5-aminocyclohexa-1,3-diene-1-carboxylic acid;hydrochloride
SMIL Cl.NC1CC(=CC=C1)C(=O)O
Tekniske data
Kemisk navn eller materiale D,L-Gabaculine, Hydrochloride
Renhedsgrad noter HPLC
InChI formel InChI=1S/C7H9NO2.ClH/c8-6-3-1-2-5(4-6)7(9)10;/h1-3,6H,4,8H2,(H,9,10);1H
Mængde 250 mg
Formel vægt 175.04
Procent renhed >95%