Få mere at vide
D,L-Cystathionine-d4, TRC
Artikelnummer. 30360769
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30360769 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30360769 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98949710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D,L-Cystathionine-d4
CAS 146764-57-0
Renhedsgrad noter HPLC
Molekylær formel C7 2H4 H10 N2 O4 S
Mængde 10 mg
Synonym Homocysteine-3,3,4,4-d4, S-(2-amino-2-carboxyethyl)- (9CI), DL-Homocysteine-3,3,4,4-d4, S-(2-amino-2-carboxyethyl)- (ZCI), DL-Cystathionine-d4, S-[(2RS)-2-Amino-2-carboxyethyl]-DL-homocysteine-3,3,4,4-d4, 2-Amino-4-[(2-amino-2-carboxyethyl)thio]butanoic-3,3,4,4-d4 acid, S-(2-Amino-2-carboxyethyl)-DL-homocysteine-3,3,4,4-D4
InChI formel InChI=1S/C7H14N2O4S/c8-4(5(10)11)3-7(9,1-2-14)6(12)13/h4,14H,1-3,8-9H2,(H,10,11)(H,12,13)/i1D2,2D2
SMIL [2H]C([2H])(S)C([2H])([2H])C(N)(CC(N)C(=O)O)C(=O)O
IUPAC navn 2,4-diamino-2-(1,1,2,2-tetradeuterio-2-sulfanylethyl)pentanedioic acid
Molekylvægt (g/mol) 226.29
Formel vægt 226.0925
Procent renhed >95