Få mere at vide
D,L Carnitine-d9 Chloride, TRC
Artikelnummer. 30350155
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30350155 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350155 Leverandør Toronto Research Chemicals Leverandørnr. TRCC18410210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D,L Carnitine-d9 Chloride
CAS 1219386-75-0
Molekylær formel C7 D9 H7 N O3 . Cl
Mængde 10 mg
Synonym 3-Carboxy-2-hydroxy-N,N,N-(trimethyl-d9)-1-propanaminium Chloride, (3-Carboxy-2-hydroxypropyl)(trimethyl-d9)ammonium Chloride, Aplegin-d9, Bicarnesine-d9, (+/-)-Carnitine Chloride
InChI formel InChI=1S/C7H15NO3.ClH/c1-8(2,3)5-6(9)4-7(10)11;/h6,9H,4-5H2,1-3H3;1H/i1D3,2D3,3D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](CC(O)CC(=O)O)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn (3-carboxy-2-hydroxypropyl)-tris(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 206.72
Formel vægt 206.1384