Få mere at vide
D,L- Carbidopa-d5, TRC
Artikelnummer. 30361243
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361243 10 mg
30358325 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30361243 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17591710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1185134-76-2
Molekylær formel C10 D5 H9 N2 O4
Molekylvægt (g/mol) 231.26
Synonym α-Hydrazino-3,4-dihydroxy-α-methyl-benzenepropanoic Acid-d5, DL-α-Hydrazino-3,4-dihydroxy-α-methylhydrocinnamic Acid-d5, α-Hydrazino-α-methyldopa-d5, dl-MK 485-d5, NSC 92521-d5
IUPAC navn 3,3,3-trideuterio-2-[dideuterio-(3,4-dihydroxyphenyl)methyl]-2-hydrazino-propanoic acid
SMIL [2H]C([2H])([2H])C(NN)(C(=O)O)C([2H])([2H])c1ccc(O)c(O)c1
Tekniske data
Kemisk navn eller materiale Carbidopa-d5
Mængde 10 mg
InChI formel InChI=1S/C10H14N2O4/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6/h2-4,12-14H,5,11H2,1H3,(H,15,16)/i1D3,5D2
Formel vægt 231.1267