missing translation for 'onlineSavingsMsg'
Få mere at vide

D,L-2-Amino-3-(hydroxy-15N-amino)propionic Acid, TRC

Artikelnummer. 30315568
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30315568 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30315568 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6111921MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale D,L-2-Amino-3-(hydroxy-15N-amino)propionic Acid
CAS 1219426-14-8
Molekylær formel C3H8N15NO3
Mængde 1 mg
Synonym alpha-Amino-beta-hydroxylaminopropionic Acid-15N, 3-(Hydroxyamino)alanine-15N, NSC 159165-15N
InChI formel InChI=1S/C3H8N2O3/c4-2(1-5-8)3(6)7/h2,5,8H,1,4H2,(H,6,7)/i5+1
SMIL NC(C[15NH]O)C(=O)O
IUPAC navn 2-amino-3-(hydroxyamino)propanoic acid
Molekylvægt (g/mol) 121.1
Formel vægt 121.0505
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.