missing translation for 'onlineSavingsMsg'
Få mere at vide

D-Homoarginine Hemisulfate, TRC

Artikelnummer. 30395586
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30395586 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30395586 Leverandør Toronto Research Chemicals Leverandørnr. TRCH585005500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale D-Homoarginine Hemisulfate
CAS 32733-73-6
Molekylær formel 2(C7H16N4O2) • (H2O4S)
Mængde 500 mg
Synonym N6-(Aminoiminomethyl)-D-lysine Hemisulfate Salt, (R)-2-Amino-6-guanidinohexanoic Acid Hemisulfate Salt
InChI formel InChI=1S/C7H16N4O2.H2O4S/c8-5(6(12)13)3-1-2-4-11-7(9)10;1-5(2,3)4/h5H,1-4,8H2,(H,12,13)(H4,9,10,11);(H2,1,2,3,4)/t5-;/m1./s1
SMIL N[C@H](CCCCNC(N)=N)C(O)=O.O=S(O)(O)=O
Molekylvægt (g/mol) 2188.239808
Formel vægt 474.222
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.