Få mere at vide
D-Homoanalog of Triamcinolone, TRC
Artikelnummer. 30132705
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132705 100 mg
30136524 50 mg
30010357 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132705 Leverandør Toronto Research Chemicals Leverandørnr. TRCH585010100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 130063-07-9
Molekylær formel C21H27FO6
Molekylvægt (g/mol) 394.43
Synonym (11beta,16alpha,17alpha)-9-Fluoro-11,16,17-trihydroxy-17-(hydroxymethyl)-D-homoandrosta-1,4-diene-3,17a-dione, (2R,3R,4aS,4bS,10aS,10bR,11S,12aS)-10b-Fluoro-3,4,4a,5,6,10a,10b,11,12,12a-decahydro-2,3,11-trihydroxy-2-(hydroxymethyl)-10a,12a-dimethyl-1,8(2H,4bH)-chrysenedione;
IUPAC navn (2R,3R,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-2,3,11-trihydroxy-2-(hydroxymethyl)-10a,12a-dimethyl-3,4,4a,4b,5,6,11,12-octahydrochrysene-1,8-dione
SMIL C[C@]12C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@](O)(CO)C2=O
Tekniske data
Kemisk navn eller materiale D-Homoanalog of Triamcinolone
Mængde 100 mg
InChI formel InChI=1S/C21H27FO6/c1-18-9-16(26)21(22)13(4-3-11-7-12(24)5-6-19(11,21)2)14(18)8-15(25)20(28,10-23)17(18)27/h5-7,13-16,23,25-26,28H,3-4,8-10H2,1-2H3/t13-,14-,15+,16-,18-,19-,20+,21-/m0/s1
Formel vægt 394.1792