missing translation for 'onlineSavingsMsg'
Learn More

D-Glyceraldehyde (>80%), TRC

Product Code. 30137016
Change view
Click to view available options
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30137016 250 mg
30135004 1 g
30008781 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137016 Supplier Toronto Research Chemicals Supplier No. TRCG598200250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 453-17-8
Molekylær formel C3 H6 O3
Molekylvægt (g/mol) 90.08
Synonym Propanal, 2,3-dihydroxy-, (2R)-, Glyceraldehyde, D- (8CI), Propanal, 2,3-dihydroxy-, (R)-, (2R)-2,3-Dihydroxypropanal, (+)-Glyceraldehyde, (2R)-2,3-Dihydroxypropanal, (R)-(+)-Glyceraldehyde, (R)-2,3-Dihydroxypropanal, (R)-Glyceraldehyde, D-(+)-Glyceraldehyde, D-Glyceraldehyde, D-Glycerose, NSC 91534, Triose
IUPAC navn (2R)-2,3-dihydroxypropanal
SMIL OC[C@@H](O)C=O

Specifications

Kemisk navn eller materiale D-Glyceraldehyde
Mængde 250 mg
InChI formel InChI=1S/C3H6O3/c4-1-3(6)2-5/h1,3,5-6H,2H2/t3-/m0/s1
Formel vægt 90.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.