Få mere at vide
D-Glucose-6,6-d2, CDN
Artikelnummer. 30318756
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30318756 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318756 Leverandør CDN Leverandørnr. CDND09001G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Glucose-6,6-d2
CAS 18991-62-3
Molekylær formel C6 2H2 H10 O6
Mængde 1 g
Synonym D-Glucose-6,6-C-d2, D-Glucose-6,6-d2 (8 CI), 6,6-Dideuterioglucose, D-Glucose-6,6-2H2, [6-2H2]-D-Glucose
InChI formel InChI=1 S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12 H,2H2/t3-,4+,5+,6+/m0/s1/i2D2
SMIL [2 H]C([2 H])(O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O
IUPAC navn (2 R,3 S,4 R,5 R)-6,6-dideuterio-2,3,4,5,6-pentahydroxyhexanal
Molekylvægt (g/mol) 182.17
Formel vægt 182.0759 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity