Få mere at vide
D-Glucose-13C6, TRC
Artikelnummer. 30004113
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004113 250 mg
30012140 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004113 Leverandør Toronto Research Chemicals Leverandørnr. TRCG595002250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110187-42-3
Molekylær formel 13C6 H12 O6
Molekylvægt (g/mol) 186.11
Synonym D-Glucopyranose, Dextrose, D-GLC, Labeled glucose
IUPAC navn (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxy(1,2,3,4,5,6-^{13}C_{6})hexanal
SMIL O[13CH2][13C@@H]1O[13CH](O)[13C@@H](O)[13C@@H](O)[13C@@H]1O
Tekniske data
Kemisk navn eller materiale D-Glucose 13C6
Renhedsgrad noter HPLC
Mængde 250 mg
InChI formel InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4-,5-,6?/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 186.084
Procent renhed >95%