Få mere at vide
D-Glucitol-d8, TRC
Artikelnummer. 30240732
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240732 10 mg
30240545 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30240732 Leverandør Toronto Research Chemicals Leverandørnr. TRCG41582210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 287962-59-8
Molekylær formel C6 D8 H6 O6
Molekylvægt (g/mol) 190.22
Synonym Cordycepic Acid-d8, Brightmoon-d8, Bronchitol-d8, D-(-)-Mannitol-d8, D-Mannitol-d8, D-Mannit-d8, Diosmol-d8, Isotol-d8, Manicol-d8, Maniton S-d8, Mannidex-d8, Mannigen-d8, Mannistol-d8, Mannit-d8, Mannit 60-d8, Mannit P-d8, Mannit S-d8, Mannite-d8, Mannitol-d8, Mannitol 35-d8, Mannitol 60-d8, Mannitol P 60-d8, Mannitolum-d8, Mannogem 2080-d8, Mannogem EZ-d8, Marine Crystal-d8, Nonpareil 108-d8, Orocell 200-d8, Osmitrol-d8, Osmosal-d8, Parteck 300-d8, Parteck Delta M-d8, Parteck M 100-d8, Parteck M 200-d8, Parteck M 300-d8, Partek M-d8, Pearlitol-d8, Perteck M-d8, Resectisol-d8, D-Sorbitol-d8, Glucarinev-d8, Esasorb-d8, Cholaxine-d8, Karion-d8, Sionite-d8, Sionon-d8
IUPAC navn (2R,3R,4R,5R)-1,1,2,3,4,5,6,6-octadeuteriohexane-1,2,3,4,5,6-hexol
SMIL [2H]C([2H])(O)[C@@]([2H])(O)[C@@]([2H])(O)[C@]([2H])(O)[C@]([2H])(O)C([2H])([2H])O
Tekniske data
Kemisk navn eller materiale D-Glucitol-d8
Mængde 10 mg
InChI formel InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1/i1D2,2D2,3D,4D,5D,6D
Formel vægt 190.1293