Få mere at vide
D-Galactose-6-O-sulphate Sodium Salt, TRC
Artikelnummer. 30179306
Skift visning
Mængde:
1 mg
2 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179306 2 mg
30179381 1 mg
30179554 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179306 Leverandør Toronto Research Chemicals Leverandørnr. TRCG1552982MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 125455-62-1
Molekylær formel C6H11O9SNa
Molekylvægt (g/mol) 282.2
Synonym D-Galactose 6-(Hydrogen Sulfate) Sodium Salt (1:1)
IUPAC navn sodium;[(2R,3R,4S,5R)-3,4,5,6-tetrahydroxyoxan-2-yl]methyl sulfate
SMIL [Na+].OC1O[C@H](COS(=O)(=O)[O-])[C@H](O)[C@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale D-Galactose-6-O-sulphate Sodium Salt
Mængde 2 mg
InChI formel InChI=1S/C6H12O9S.Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;/h2-10H,1H2,(H,11,12,13);/q;+1/p-1/t2-,3+,4+,5-,6?;/m1./s1
Formel vægt 282.0021