Få mere at vide
D-Fructose-4,5,6,6-d4, TRC
Artikelnummer. 30403301
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30403301 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403301 Leverandør Toronto Research Chemicals Leverandørnr. TRCF79255425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Fructose-4,5,6,6-d4
CAS 478518-49-9
Molekylær formel C6H8D4O6
Mængde 25 mg
Synonym Advantose FS 95-4,5,6-d4, D-(-)-Fructose-4,5,6-d4, D-(-)-Levulose-4,5,6-d4, D-Arabino-2-hexulose-4,5,6-d4, Fructose-4,5,6-d4, Fruit Sugar-4,5,6-d4, Fujifructo L 95-4,5,6-d4, Furucton-4,5,6-d4, Hi-Fructo 970-4,5,6-d4, Krystar-4,5,6-d4, Krystar 300v, Levulose-4,5,6-d4, Nevulose-4,5,6-d4, Sugar Fruit-4,5,6-d4
InChI formel InChI=1S/C6H12O6/c7-2-6(11)5(10)4(9)3(8)1-12-6/h3-5,7-11H,1-2H2
SMIL O[C@]1([2H])[C@]([C@@H](O)[C@@](CO)(O)OC1([2H])[2H])([2H])O
Molekylvægt (g/mol) 184.18