Få mere at vide
D-Fructose-13C6, TRC
Artikelnummer. 30137976
Skift visning
Mængde:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137976 25 mg
17999984 100 mg
17918494 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137976 Leverandør Toronto Research Chemicals Leverandørnr. TRCF79254625MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 201595-65-5
Molekylær formel 13C6 H12 O6
Molekylvægt (g/mol) 186.11
Synonym D-Fructose-1,2,3,4,5,6-13C6, D-Fructose-13C6 (9CI), D-[1,2,3,4,5,6-13C6]Fructose, D-[13C6]Fructose
IUPAC navn (3S,4R,5R)-1,3,4,5,6-pentahydroxy(1,2,3,4,5,6-^{13}C_{6})hexan-2-one
SMIL O[13CH2][13C@@H](O)[13C@@H](O)[13C@H](O)[13C](=O)[13CH2]O
Tekniske data
Kemisk navn eller materiale D-Fructose-13C6
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3,5-9,11-12H,1-2H2/t3-,5-,6-/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 186.0835
Procent renhed >95%