missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-1,6-diphosphate dicalcium salt, 95%

Catalog Number p-7043240
Click to view available options
Quantity:
25 g
100 g
250 g
This item is not returnable. View return policy
6055-82-9
C6H10Ca2O12P2
416.238
MFCD00135875
MKSZTTCFYSGQHA-UHFFFAOYSA-J
d-fructose, 1,6-bis dihydrogen phosphate , calcium salt 1:2, dicalcium 2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl phosphate, dicalcium 2,3,4-tris oxidanyl-5-oxidanylidene-6-phosphonatooxy-hexyl phosphate
53398690
dicalcium;(2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl) phosphate
C(C(C(C(C(=O)COP(=O)([O-])[O-])O)O)O)OP(=O)([O-])[O-].[Ca+2].[Ca+2]
This item is not returnable. View return policy
6055-82-9
C6H10Ca2O12P2
416.238
MFCD00135875
MKSZTTCFYSGQHA-UHFFFAOYSA-J
d-fructose, 1,6-bis dihydrogen phosphate , calcium salt 1:2, dicalcium 2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl phosphate, dicalcium 2,3,4-tris oxidanyl-5-oxidanylidene-6-phosphonatooxy-hexyl phosphate
53398690
dicalcium;(2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl) phosphate
C(C(C(C(C(=O)COP(=O)([O-])[O-])O)O)O)OP(=O)([O-])[O-].[Ca+2].[Ca+2]

D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

Solubility
Soluble in water

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY
Color White
Quantity 25 g
Solubility Information Soluble in water
Formula Weight 418.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-Fructose-1,6-diphosphate dicalcium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'