Få mere at vide
D-DOPA, TRC
Artikelnummer. 17999454
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17999454 100 mg
17959744 250 mg
17999354 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17999454 Leverandør Toronto Research Chemicals Leverandørnr. TRCD533750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5796-17-8
Molekylær formel C9H11NO4
Molekylvægt (g/mol) 197.19
Synonym D-Tyrosine, 3-hydroxy-, Alanine, 3-(3,4-dihydroxyphenyl)-, D- (8CI), 3-Hydroxy-D-tyrosine, (+)-3-(3,4-Dihydroxyphenyl)alanine, (+)-Dopa, 3,4-Dihydroxy-D-phenylalanine, 3-(3,4-Dihydroxyphenyl)-D-alanine, D-3,4-Dihydroxyphenylalanine, D-3-(3,4-Dihydroxyphenyl)alanine, D-3-Hydroxytyrosine, D-Dopa, β-(3,4-Dihydroxy)-D-phenylalanine, (2R)-2-Amino-3-(3,4-dihydroxyphenyl)propanoic Acid
SMIL N[C@H](Cc1ccc(O)c(O)c1)C(=O)O
Tekniske data
Kemisk navn eller materiale D-DOPA
Renhedsgrad noter HPLC
InChI formel InChI=1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m1/s1
Mængde 100 mg
Formel vægt 197.0688
Procent renhed >95%