Få mere at vide
D-Cysteine Methyl Ester Hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | D-Cysteine Methyl Ester Hydrochloride |
| CAS | 70361-61-4 |
| Molekylær formel | C4H10ClNO2S |
| Mængde | 2.5 g |
| Synonym | (S)-Cysteine Methyl Ester Hydrochloride, Methyl D-Cysteinate Hydrochloride |
| InChI formel | InChI=1S/C4H9NO2S.ClH/c1-7-4(6)3(5)2-8;/h3,8H,2,5H2,1H3;1H/t3-;/m1./s1 |
| SMIL | N[C@H](CS)C(OC)=O.Cl |
| IUPAC navn | methyl D-cysteinate hydrochloride |
| Molekylvægt (g/mol) | 171.65 |
| Formel vægt | 171.01 |