Få mere at vide
D-Cysteine Ethyl Ester Hydrochloride, TRC
Artikelnummer. 30179437
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179437 1 g
30179342 250 mg
30179065 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179437 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9950451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75521-14-1
Molekylær formel C5H11NO2S . HCl
Molekylvægt (g/mol) 185.67
Synonym Ethyl D-cysteinate Hydrochloride;
IUPAC navn ethyl (2S)-2-amino-3-sulfanylpropanoate;hydrochloride
SMIL Cl.CCOC(=O)[C@H](N)CS
Tekniske data
Kemisk navn eller materiale D-Cysteine Ethyl Ester Hydrochloride
InChI formel InChI=1S/C5H11NO2S.ClH/c1-2-8-5(7)4(6)3-9;/h4,9H,2-3,6H2,1H3;1H/t4-;/m1./s1
Mængde 1 g
Formel vægt 185.0277