Learn More
Alfa Aesar™ D-(+)-Cellopentaose, 95%
Brand:  Alfa Aesar™ J67561.8+
Additional Details : Weight : 0.00002kg
Quantity 2mg
 View more versions of this product
Product Code. 15478759
  • 1148.53 DKK / 2mg
Estimated Shipment
26-10-2020
Add to basket
Description
Description
  • Used by cellulosic bacteria as an energy source
  • Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidase(s)
  • Can also be used in saccharification and ethano
Specifications
Chemical Identifiers
2240-27-9
MFCD09750881
Cellopentaose
Specifications
D-(+)-Cellopentaose
White
2240-27-9
79497
828.72
95%
Hygroscopic
2mg
MFCD09750881
Cellopentaose
Powder
Videos
SDS Documents