missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Cellopentaose, 95%, Thermo Scientific Chemicals

Catalog Number p-7057920
Click to view available options
Quantity:
1 mg
2 mg
5 mg
This item is not returnable. View return policy
2240-27-9
MFCD09750881
Cellopentaose
This item is not returnable. View return policy
2240-27-9
MFCD09750881
Cellopentaose

Used by cellulosic bacteria as an energy source. Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidase(s). Can also be used in saccharification and ethanol fermentation research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Used by cellulosic bacteria as an energy source
  • Used to identify, differentiate and characterize oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidase(s)
  • Can also be used in saccharification and ethano
TRUSTED_SUSTAINABILITY
Color White
Quantity 1 mg
Beilstein 79497
Sensitivity Hygroscopic
Formula Weight 828.72
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-(+)-Cellopentaose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'