missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ D-Aspartic acid 1-benzyl ester, 98%

Brand:  Alfa Aesar™ H63362.06

Additional Details : CAS Number : 79337-40-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470019

  • 3066.18 DKK / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

D-Aspartic acid 1-benzyl ester
223.23
H-D-Asp-OBzl
79337-40-9
MFCD00037289

Specifications

D-Aspartic acid 1-benzyl ester
Solid
223.23
H-D-Asp-OBzl
79337-40-9
98%
MFCD00037289
5g
Videos
SDS