missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-(+)-Arabitol, 99%
Click to view available options
Quantity:
5 g
25 g
Description
It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.
Solubility
Soluble in water (50 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
It can be employed as a sugar in the production of xylitol via biotransformation process. It is a substrate used to identify, differentiate and characterize enzyme such as the gluconobacter oxydans dehydrogenase(s), Gox2181, hyperthermophilic D-arabitol dehydrogenase from Thermotoga maritime and NAD-dependent D-arabitoldehydrogenase from acetic acid bacterium, Acetobacter suboxydans.
Solubility
Soluble in water (50 mg/ml).
Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
Specifications
Melting Point | 100°C to 104°C |
Quantity | 5 g |
Beilstein | 1720520 |
Merck Index | 14,762 |
Solubility Information | Soluble in water (50mg/ml). |
Optical Rotation | +132° (c=0.4 in Acidified Molybdate Solution) |
Formula Weight | 152.15 |
Percent Purity | 99% |
Chemical Name or Material | D-(+)-Arabitol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'