Få mere at vide
D-Arabinose-5-13C, TRC
Artikelnummer. 30136792
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136792 100 mg
30008581 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136792 Leverandør Toronto Research Chemicals Leverandørnr. TRCA764179100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 139657-60-6
Molekylær formel C413CH10O5
Molekylvægt (g/mol) 151.12
Synonym D-[5-13C]Arabinose
IUPAC navn 2,3,4,5-tetrahydroxypentanal
SMIL [H][13C]1([C@@H](O)[C@@H](O)[C@H](O)C(O)O1)[H]
Tekniske data
Kemisk navn eller materiale D-Arabinose-5-13C
Mængde 100 mg
InChI formel InChI=1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2
Formel vægt 151.0562