Få mere at vide
D-Allose-13C, TRC
Artikelnummer. 30179034
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179034 100 mg
30179216 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179034 Leverandør Toronto Research Chemicals Leverandørnr. TRCA548006100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70849-28-4
Molekylær formel 13C C5 H11 O6
Molekylvægt (g/mol) 180.141
Synonym D-Allopyranose-13C
IUPAC navn (2R,3S,4R,5R)-2-(hydroxymethyl)-(613C)2,3,4,5-tetrahydropyran-3,4,5,6-tetrol
SMIL OC[C@H]1O[13C](O)[C@H](O)[C@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale D-Allose-13C
Mængde 100 mg
InChI formel InChI=1S/C6H11O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-5,7-11H,1H2/t2-,3-,4-,5-/m1/s1/i6+1
Formel vægt 180.059