Få mere at vide
D-Alanine-2-d1, TRC
Artikelnummer. 30316673
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316673 10 mg
30316478 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316673 Leverandør Toronto Research Chemicals Leverandørnr. TRCA48100110MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Alanine-2-D
CAS 56595-54-1
Molekylær formel C3 D H6 N O2
Mængde 10 mg
Synonym D-Alanine-2-d (9CI, ACI), (2R)-2-Amino-2-deuteriopropanoic acid, D-(α2H)Alanine, (2R)-2-Aminopropanoic-2-d acid, D-Alanine-2-D, D-[2-2H1]Alanine
InChI formel InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1/i2D
SMIL [2H][C@](C)(N)C(=O)O
IUPAC navn (2R)-2-amino-2-deuteriopropanoic acid
Molekylvægt (g/mol) 90.0993
Formel vægt 90.054