missing translation for 'onlineSavingsMsg'
Learn More

D-64131, Thermo Scientific™

Catalog Number 15437179
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Novel inhibitor of tubulin polymerization

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 74588-78-6
Color Yellow
Molecular Formula C16H13NO2
MDL Number MFCD04039791
Quantity 10 mg
Synonym 5-methoxy-1h-indol-2-yl phenylmethanone, 5-methoxy-1h-indol-2-yl-phenylmethanone, 5-methoxy-1h-indol-2-yl phenyl methanone, 5-methoxy-1h-indol-2-yl-phenyl-methanone, 2-benzoyl-5-methoxy-1h-indole, tocris-1643, 5-methoxy-2-benzoylindole, 2-benzoyl-5-methoxyindole
InChI Key ICMIJSRDISNKOC-UHFFFAOYSA-N
SMILES COC1=CC=C2NC(=CC2=C1)C(=O)C1=CC=CC=C1
IUPAC Name (5-methoxy-1H-indol-2-yl)-phenylmethanone
Molecular Weight (g/mol) 251.29
PubChem CID 3921152
Formula Weight 251.28
Physical Form Crystalline Solid
Chemical Name or Material D-64131
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'