Få mere at vide
D-4-Hydroxyphenylalanine-3,3-d2, CDN
Artikelnummer. 30328028
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328028 0.25 g -
30312663 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328028 Leverandør CDN Leverandørnr. CDND74870.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Tyrosine-d2
CAS 1202064-22-9
Molekylær formel C9 D2 H9 N O3
Mængde 0.25 g
Synonym D-Tyrosine-β,β-d2 (ACI), D-Tyrosine-3,3-d2, D-(β,β-2H2)Tyrosine, D-Tyrosine-D2 (propanoic-3,3-D2 acid), (2 R)-2-Amino-3-(4-hydroxyphenyl)propanoic-3,3-d2 acid, 4-Hydroxyphenyl-D-alanine-3,3-d2, β-(p-Hydroxyphenyl)-D-alanine-β,β-d2, D-Tyrosine-d2, D-4-Hydroxyphenylalanine-3,3-d2
InChI formel InChI=1 S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11 H,5,10H2,(H,12,13)/t8-/m1/s1/i5D2
SMIL [2 H]C([2 H])([C@@H](N)C(=O)O)c1ccc(O)cc1
IUPAC navn (2 R)-2-amino-3,3-dideuterio-3-(4-hydroxyphenyl)propanoic acid
Molekylvægt (g/mol) 183.201
Formel vægt 183.086 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity