Få mere at vide
D-2-Aminohexanoic-3,3,4,4,5,5,6,6,6-d9 Acid, CDN
Artikelnummer. 30320847
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320847 0.005 g
30316365 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320847 Leverandør CDN Leverandørnr. CDND79590.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Norleucine-3,3,4,4,5,5,6,6,6-D9
CAS 2185812-01-3
Molekylær formel C6 D9 H4 N O2
Mængde 0.005 g
Synonym (2 R)-2-Amino-3,3,4,4,5,5,6,6,6-nonadeuteriohexanoic acid, D-Norleucine-d9, D-Norleucine-3,3,4,4,5,5,6,6,6-D9, D-2-Aminohexanoic-3,3,4,4,5,5,6,6,6-d9 Acid
InChI formel InChI=1 S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m1/s1/i1D3,2D2,3D2,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[C@@H](N)C(=O)O
IUPAC navn (2 R)-2-amino-3,3,4,4,5,5,6,6,6-nonadeuteriohexanoic acid
Molekylvægt (g/mol) 140.2284
Formel vægt 140.1511 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity