Få mere at vide
D-2-Aminobutyric-2,3,3,4,4,4-d6 Acid, TRC
Artikelnummer. 30309160
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309160 1 mg
30321898 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309160 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6029161MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-2-Aminobutanoic-2,3,3,4,4,4-D6 Acid
CAS 1276197-57-9
Molekylær formel C4 D6 H3 N O2
Mængde 1 mg
Synonym (2R)-2-Aminobutanoic-2,3,3,4,4,4-d6 acid, D-2-Aminobutyric acid-d6, D-Butyrine-d6, D-Ethylglycine-d6, D-Homoalanine-d6, D-α-Aminobutyric acid-d6, (R)-2-Aminobutanoic acid-d6, (R)-2-Aminobutanoic-2,3,3,4,4,4-D6 acid
InChI formel InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m1/s1/i1D3,2D2,3D
SMIL [2H]C([2H])([2H])C([2H])([2H])[C@@]([2H])(N)C(=O)O
IUPAC navn (2R)-2-amino-2,3,3,4,4,4-hexadeuteriobutanoic acid
Molekylvægt (g/mol) 109.157
Formel vægt 109.101