Få mere at vide
D-2-Aminobutyric-2,3,3,4,4,4-d6 Acid, CDN
Artikelnummer. 30322232
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322232 0.1 g
30318161 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322232 Leverandør CDN Leverandørnr. CDND70120.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (R)-2-Aminobutanoic-2,3,3,4,4,4-D6 Acid
CAS 1276197-57-9
Molekylær formel C4 D6 H3 N O2
Mængde 0.1 g
Synonym (2 R)-2-Aminobutanoic-2,3,3,4,4,4-d6 acid, D-2-Aminobutyric acid-d6, D-Butyrine-d6, D-Ethylglycine-d6, D-Homoalanine-d6, D-α-Aminobutyric acid-d6, (R)-2-Aminobutanoic acid-d6, (R)-2-Aminobutanoic-2,3,3,4,4,4-D6 acid
InChI formel InChI=1 S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m1/s1/i1D3,2D2,3 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])[C@@]([2 H])(N)C(=O)O
IUPAC navn (2 R)-2-amino-2,3,3,4,4,4-hexadeuteriobutanoic acid
Molekylvægt (g/mol) 109.157
Formel vægt 109.101 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity