missing translation for 'onlineSavingsMsg'
Learn More

D(+)-2-Amino-3-phenyl-1-propanol, 98%, Thermo Scientific Chemicals

Catalog Number p-58652
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
5267-64-1
C9H13NO
151.21
MFCD00064298
d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol
This item is not returnable. View return policy
5267-64-1
C9H13NO
151.21
MFCD00064298
d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 90°C to 94°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 23
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,benzene,chloroform
Formula Weight 151.21
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material D(+)-2-Amino-3-phenyl-1-propanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • D(+)-2-Amino-3-phenyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'