Få mere at vide
D 13223-d4 (Flupirtine Metabolite), TRC
Artikelnummer. 30349644
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30349644 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30349644 Leverandør Toronto Research Chemicals Leverandørnr. TRCD10006725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D 13223-d4 (Flupirtine Metabolite)
CAS 1216989-17-1
Molekylær formel C14H12D4ClFN4O
Mængde 25 mg
Synonym N-[2-Amino-6-[[(4-fluorophenyl-d4)methyl]amino]-3-pyridinyl]acetamide Hydrochloride
InChI formel InChI=1S/C14H15FN4O.ClH/c1-9(20)18-12-6-7-13(19-14(12)16)17-8-10-2-4-11(15)5-3-10;/h2-7H,8H2,1H3,(H,18,20)(H3,16,17,19);1H/i2D,3D,4D,5D;
SMIL FC1=C([2H])C([2H])=C(CNC2=CC=C(NC(C)=O)C(N)=N2)C([2H])=C1[2H].Cl
IUPAC navn N-[2-amino-6-[(2,3,5,6-tetradeuterio-4-fluorophenyl)methylamino]pyridin-3-yl]acetamide;hydrochloride
Molekylvægt (g/mol) 314.78