missing translation for 'onlineSavingsMsg'
Learn More

D(+)-10-Camphorsulfonic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-82134
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
3144-16-9
C10H15O4S
231.29
MFCD00064157,MFCD00074827
MIOPJNTWMNEORI-XVKPBYJWSA-M
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
65617
CHEBI:55403
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2
3144-16-9
C10H15O4S
231.29
MFCD00064157,MFCD00074827
MIOPJNTWMNEORI-XVKPBYJWSA-M
1s,4r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonic acid, d-camphorsulfonic acid, r-camphorsulfonic acid, unii-9tlz01s15l, d-+-10-camphorsulfonic acid, d-+-camphor-10-sulfonic acid, dl-10-camphorsulfonic acid, camphor-10-sulfonic acid, camphersulfosaeure german, d-camphor-10-sulfonic acid
65617
CHEBI:55403
[(1R,4S)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
CC1(C)[C@H]2CC[C@]1(CS([O-])(=O)=O)C(=O)C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >190.0°C
Color White
Loss on Drying 2% max. (1 g, 105°C, 5 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 11,IV,642
Fieser 01,108; 02,58; 04,68; 13,62; 14,71; 16,58
Merck Index 15, 1736
Specific Rotation + 22.00
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in glacial acetic acid,slightly soluble in ethyl acetate,practically insoluble in ether,very soluble in ethanol
Formula Weight 232.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-10-Camphorsulfonic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • D(+)-10-Camphorsulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'