Få mere at vide
Tocris Bioscience™ CZC 25146, Tocris Bioscience™
GREENER_CHOICE
Potent LRRK2 inhibitor
Brand:  Tocris Bioscience™ 6071/50
Encompass
Produktkode 15737206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
CZC 25146 is a potent LRRK2 inhibitor (IC50 values are 4.76 and 6.87 nM for for wild-type and G2019S mutant forms of LRRK2, respectively). Also inhibits PLK4, GAK, TNK1, CAMKK2 and PIP4K2. Attenuates mutant LRRK2-induced injury of cultured rodent and human neurons. Cell permeable.
Tekniske data
Tekniske data
CZC 25146
N-[2-[[5-Fluoro-2-[[2-methoxy-4-(4-morpholinyl)phenyl]amino]-4-pyrimidinyl]amino]phenyl]methanesulfonamide
Soluble to 100mM in DMSO
Pale Purple
1191911-26-8
50 mg
Solid
Bioactive Small Molecule
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only.