Learn More
Novus Biologicals™ CyTRAK Orange™
Product Code. 18054234 Shop All Bio Techne Products
Click to view available options
Mængde:
50 μL
200 μL
This item is not returnable. View return policy
Product Code. 18054234
Brand: Novus Biologicals™ NBP28112750UL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This item is not returnable. View return policy
Specifications
Indhold og opbevaring Store at 4°C in the dark. Do not freeze.
Mængde 50 μL
Til brug med (applikation) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy